missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566488
This item is not returnable.
View return policy
Description
Immunogen sequence: LEPVNFPSHFL LRWCQGAEGA TLDIFDYIYI DAFGKGKQLT VKECEYLIGQ HVTAALYGVV NVKKVLQ Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternative splicing of this gene generates 2 transcript variants.
Specifications
| FBXO21 | |
| Polyclonal | |
| Unconjugated | |
| FBXO21 | |
| 2810425J22Rik; AU016673; F-box only protein 21; F-box protein 21; FBX21; FBXO21; KIAA0875; mKIAA0875 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23014 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94952 | |
| FBXO21 | |
| Recombinant Human FBXO21. Recombinant protein control fragment (Product #RP-107161). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction