missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523718
This item is not returnable.
View return policy
Description
FBXO2 Polyclonal Antibody for Western Blot, Flow
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. This protein is highly similar to the rat NFB42 (neural F Box 42 kDa) protein which is enriched in the nervous system and may play a role in maintaining neurons in a postmitotic state.
Specifications
| FBXO2 | |
| Polyclonal | |
| Unconjugated | |
| FBXO2 | |
| FBG1; F-box gene 1; F-box only protein 2; F-box protein 2; Fbs1; Fbs2; FBX2; FBXO2; neural F box protein NFB42; NFB42; OCP1; organ of Corti protein 1; prion protein ligand 4; Prpl4 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 230904, 26232 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q80UW2, Q9UK22 | |
| FBXO2 | |
| KLH conjugated synthetic peptide between 244-271 amino acids from the C-terminal region of human FBXO2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction