Learn More
FBXO17 Mouse, anti-Human, CoraLite Plus 488, Clone: 1F3D3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68246-20UL
Description
The FBXO17 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FBXO17. This antibody recognizes human antigen. The FBXO17 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| FBXO17 | |
| 1F3D3 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| F box only protein 17, F box only protein 26, F box protein 17, FBG4, Fbx17, FBX26, FBXO17, FBXO26, FLJ11798, FLJ25205, FLJ58300 | |
| 20 μL | |
| 115290 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |