Learn More
FBXO11 Mouse anti-Human, Unconjugated, Clone: 3C5E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67365-1-IG-20UL
This item is not returnable.
View return policy
Description
The FBXO11 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FBXO11. This antibody recognizes human antigen. The FBXO11 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| FBXO11 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 3C5E1 | |
| Unconjugated | |
| 20 μL | |
| 80204 | |
| IgG1 |