Learn More
FBXO11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand:  Novus Biologicals NB100-59826
Product Code. 18701534
  • £468.00 / 0.10mL
Description
FBXO11 Polyclonal specifically detects FBXO11 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Knockout Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunocytochemistry, Immunofluorescence
Western Blot 1:2000-1:10000, Simple Western 1:40, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 2 - 5 ug/mg lysate, Knockout Validated
FBXO11