missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXO10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113506
This item is not returnable.
View return policy
Description
FBXO10 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Members of the F-box protein family, such as FBXO10, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
Specifications
| FBXO10 | |
| Polyclonal | |
| Unconjugated | |
| FBXO10 | |
| F-box only protein 10; F-box only protein 10; LOW QUALITY PROTEIN: F-box only protein 10; F-box protein 10; F-box protein Fbx10; FBX10; FBXO10; PRMT11 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 26267 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UK96 | |
| FBXO10 | |
| Recombinant Human F-box only protein 10 protein (248-400AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction