missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXL7 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98943CL1
This item is not returnable.
View return policy
Description
FBXL7 Polyclonal antibody specifically detects FBXL7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FBXL7 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 23194 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| FBL7FBL6F-box protein Fbl7, F-box and leucine-rich repeat protein 7F-box protein FBL6/FBL7, F-box/LRR-repeat protein 7, KIAA0840 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human FBXL7. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction