missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530560
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, human FBXL4-transfected 293T cells. Predicted reactivity: Mouse (91%), Rat (90%), Pig (92%), Rhesus Monkey (96%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains at least 9 tandem leucine-rich repeats.
Specifications
| FBXL4 | |
| Polyclonal | |
| Unconjugated | |
| FBXL4 | |
| AI836810; FBL4; FBL5; F-box and leucine rich repeat protein 4; F-box and leucine-rich repeat protein 4; F-box protein FBL4/FBL5; F-box/LRR-repeat protein 4; Fbxl4; MTDPS13 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26235 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.88 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9UKA2 | |
| FBXL4 | |
| Recombinant protein encompassing a sequence within the center region of human FBL4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction