missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FBXL20 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12858662
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12858662 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12858662 Supplier Invitrogen™ Supplier No. PA520486

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is A-20 cell lysate. PA5-20486 can be used with blocking peptide PEP-0606.

Members of the F-box protein family, such as Scrapper, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin, and F-box proteins, act as protein-ubiquitin ligases. Scrapper is selectively expressed in the brain, broadly expressed within the mouse CNS and is abundant at presynaptic membrane. Scrapper has orthologs in C. elegans, D. melanogaster, and mammals which suggests that it might function as an important membrane-localized E3 ligase in various species. Scrapper is a major presynaptic E3 ubiquitin ligase that acts through RIM1a via degradation and the ubiquitin-proteasome-system (UPS) pathway to critically regulate synaptic transmission. This identifies protein degradation as a mechanism for holding synaptic communication in check.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL20
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene FBXL20
Gene Accession No. Q96IG2, Q9CZV8, Q9QZH7
Gene Alias 2610511F20Rik; 4632423N09Rik; AI849362; AL117906; C86145; Fbl2; Fbl20; F-box and leucine rich repeat protein 20; F-box and leucine-rich repeat protein 20; F-box protein FBL2; F-box/LRR-repeat protein 20; F-box/LRR-repeat protein 2-like; Fbxl2; Fbxl20; mKIAA4147; Scr; scrapper
Gene Symbols FBXL20
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of the human SCRAPPER.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64039, 72194, 84961
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.