missing translation for 'onlineSavingsMsg'
Learn More

FBXL20 Antibody, FITC, Novus Biologicals™

Product Code. 18400321 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400321 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400321 Supplier Novus Biologicals Supplier No. NBP177317F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FBXL20 Polyclonal antibody specifically detects FBXL20 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL20
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias DKFZp564O2364, Fbl2, Fbl20, F-box and leucine-rich repeat protein 20FLJ21037, F-box protein FBL2, F-box/LRR-repeat protein 20, F-box/LRR-repeat protein 2-like, MGC15482
Host Species Rabbit
Immunogen Antibody was raised against a 18 amino acid synthetic peptide near the amino terminus of the human SCRAPPER. The immunogen is located within the first 50 amino acids of SCRAPPER. Amino Acid Squence: SDEAVINKKLPKELLLR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84961
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.