Learn More
FBXL19 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565146
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: TGVSKKQLMWL LNRLQGLQEL VLSGCSWLSV SALGSAPLPA LRLLDLRWIE DVKDSQLREL LLPPPDTKPG QTESRGRLQG VAELRLAGLE LTDASLRLLL RHAPQLSALD LSHCAHVGDP SVHLLT Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Substrate-recognition component of the SCF (SKP1-CUL1-F- box protein)-type E3 ubiquitin ligase complex.Specifications
FBXL19 | |
Polyclonal | |
Unconjugated | |
FBXL19 | |
6030430O11; BC059812; CXXC11; Fbl19; F-box and leucine rich repeat protein 19; F-box and leucine-rich repeat protein 19; F-box/LRR-repeat protein 19; FBXL19; JHDM1C; jumonji C domain-containing histone demethylase 1C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54620 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PCT2 | |
FBXL19 | |
Recombinant Human FBXL19 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only