missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FBXL16 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12809242
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12809242 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12809242 Supplier Invitrogen™ Supplier No. PA521094

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human spleen tissue lysate. PA5-21094 can be used with blocking peptide PEP-1208.

FBXL16 (F-box and leucine-rich repeat protein 16), a member of the F-box protein family, is a probable substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex. It is characterized by an approximately 40-amino acid F-box motif. F-box proteins interact with SKP1 through the F-box, and they interact with ubiquitination targets through other protein interaction domains. FBXL16 is a novel E2F1-regulated gene in crucial cell cycle regulation pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBXL16
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene FBXL16
Gene Accession No. A2RT62, Q5MJ12, Q8N461
Gene Alias BC042620; C16orf22; c380A1.1; c380A1.1 (novel protein); FBL16; F-box and leucine rich repeat protein 16; F-box and leucine-rich repeat protein 16; F-box/LRR-repeat protein 16; Fbxl16; Scirr1; spinal cord injury and regeneration related 1; spinal cord injury and regeneration-related protein 1
Gene Symbols FBXL16
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy terminus of human FBXL16.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146330, 214931, 494223
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.