missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXL14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100521
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FBXL14.
FBXL14 is members of the F-box protein family, such as FBXL14, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains.
Specifications
| FBXL14 | |
| Polyclonal | |
| Unconjugated | |
| Fbxl14 | |
| AW322056; FBL14; F-box and leucine rich repeat protein 14; F-box and leucine-rich repeat protein 14; F-box/LRR-repeat protein 14; Fbx14l; FBXL14; Ppa | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 101358, 144699, 312675 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BID8, Q8N1E6 | |
| Fbxl14 | |
| A synthesized peptide derived from human FBXL14(Accession Q8N1E6), corresponding to amino acid residues S81-T131. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction