Learn More
Invitrogen™ FBXL12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589921
Description
Immunogen sequence: MATLVELPDS VLLEIFSYLP VRDRIRISRV CHRWKRLVDD RWLWRHVDLT LYTMRPKVMW HLLRRYMASR LHSLRMGGYL FSGSQAPQLS PALLRALGQK CPNLKRLCLH VADLSMVPIT SLPSTLRTLE LHSCEISMAW LHKQQDPTVL PLLECIVLDR VPAFRDEHLQ GLTRFRALRS LVLGGTYRVT ETGLDAGLQE LSYLQRLEVL GCTLSADSTL LAISRHLRDV RKIRLTVRGL SAPGLAVLEG MPALESLCLQ GPLVTPEMPS PTEILSSCLT MPKLRVLELQ GLGWEGQEAE KILCKGLPHC MVIVRACPKE SMDWWM; Positive Samples: NIH/3T3, SW480, 293T, A-431, HeLa.
Members of the F-box protein family, such as FBXL12, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1.
Specifications
| FBXL12 | |
| Polyclonal | |
| Unconjugated | |
| FBXL12 | |
| 3110048D16Rik; Fbl12; F-box and leucine rich repeat protein 12; F-box and leucine-rich repeat protein 12; F-box protein containing leucine repeats 12; F-box protein FBL12; F-box/LRR-repeat protein 12; FBXL12; FLJ20188; LOC617304 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 30843, 54850 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9EPX5, Q9NXK8 | |
| FBXL12 | |
| Recombinant protein (or fragments). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.