missing translation for 'onlineSavingsMsg'
Learn More

FbxL12 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18341484 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341484 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341484 Supplier Novus Biologicals Supplier No. NBP306196G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FbxL12 Polyclonal specifically detects FbxL12 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FbxL12
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias FBL12, F-box and leucine-rich repeat protein 12FLJ20188, F-box protein FBL12, F-box/LRR-repeat protein 12
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FbxL12 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54850
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.