missing translation for 'onlineSavingsMsg'
Learn More

FBP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£360.00 - £553.00

Specifications

Antigen FBP2
Dilution Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18325306
View Documents
Novus Biologicals
NBP3-17033-25UL
25 μg
£360.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18375305
View Documents
Novus Biologicals
NBP3-17033-100UL
100 μg
£553.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FBP2 Polyclonal antibody specifically detects FBP2 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

FBP2
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Protein Phosphatase
PBS, pH 7.2, 40% glycerol
8789
IgG
Affinity purified
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Purified
RUO
Human
D-fructose-1,6-bisphosphate 1-phosphohydrolase 2, EC 3.1.3, EC 3.1.3.11, FBPase 2, fructose-1,6-bisphosphatase 2, fructose-1,6-bisphosphatase isozyme 2, hexosediphosphatase, MGC142192, muscle fructose-bisphosphatase
This antibody was developed against Recombinant Protein corresponding to amino acids: VAYIIEQAGGLATTGTQPVLDVKPEAIHQRVPLILGSPEDVQEYLTCVQKNQAGS
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.