missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565938
This item is not returnable.
View return policy
Description
Immunogen sequence: VAYIIEQAGGL ATTGTQPVLD VKPEAIHQRV PLILGSPEDV QEYLTCVQKN QAGS Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
FBP2 is a gluconeogenesis regulatory enzyme which catalyzes the hydrolysis of fructose 1,6-bisphosphate to fructose 6-phosphate and inorganic phosphate.
Specifications
| FBP2 | |
| Polyclonal | |
| Unconjugated | |
| FBP2 | |
| D-fructose-1,6-bisphosphate 1-phosphohydrolase 2; Fbp1; Fbp-1; FBP2; FBPase; FBPase 2; FBPase muscle; fructose bisphosphatase 2; fructose-1; fructose-1,6-bisphosphatase 2; fructose-1,6-bisphosphatase isozyme 2; fructose-bisphosphatase 2; hexosediphosphatase; muscle FBPase; muscle fructose-bisphosphatase; Rae-30 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8789 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00757 | |
| FBP2 | |
| Recombinant Human FBP2. Recombinant protein control fragment (Product #RP-106620). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction