missing translation for 'onlineSavingsMsg'
Learn More

FBP2 Antibody (1E11), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18420678 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420678 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420678 Supplier Novus Biologicals Supplier No. NBP222556JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FBP2 Monoclonal antibody specifically detects FBP2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FBP2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1E11
Conjugate Janelia Fluor 549
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias D-fructose-1,6-bisphosphate 1-phosphohydrolase 2, EC 3.1.3, EC 3.1.3.11, FBPase 2, fructose-1,6-bisphosphatase 2, fructose-1,6-bisphosphatase isozyme 2, hexosediphosphatase, MGC142192, muscle fructose-bisphosphatase
Host Species Mouse
Immunogen Recombinant human FBP2 (1-339aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 8789
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.