missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBP2 Antibody (1E11), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22556R
This item is not returnable.
View return policy
Description
FBP2 Monoclonal antibody specifically detects FBP2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| FBP2 | |
| Monoclonal | |
| DyLight 550 | |
| D-fructose-1,6-bisphosphate 1-phosphohydrolase 2, EC 3.1.3, EC 3.1.3.11, FBPase 2, fructose-1,6-bisphosphatase 2, fructose-1,6-bisphosphatase isozyme 2, hexosediphosphatase, MGC142192, muscle fructose-bisphosphatase | |
| Recombinant human FBP2 (1-339aa) purified from E. coli | |
| 0.1 mL | |
| Protein Phosphatase | |
| 8789 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 1E11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction