missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBP17 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27056-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the formin-binding-protein family. The protein contains an N-terminal Fer/Cdc42-interacting protein 4 (CIP4) homology (FCH) domain followed by a coiled-coil domain, a proline-rich motif, a second coiled-coil domain, a Rho family protein-binding domain (RBD), and a C-terminal SH3 domain. This protein binds sorting nexin 2 (SNX2), tankyrase (TNKS), and dynamin; an interaction between this protein and formin has not been demonstrated yet in human.Specifications
| FBP17 | |
| Polyclonal | |
| Unconjugated | |
| FNBP1 | |
| FBP17, FNBP1, formin binding protein 1, Formin binding protein 17, hFBP17, KIAA0554 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14269, 23048 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q80TY0, Q96RU3 | |
| Fnbp1 | |
| FBP17 Fusion Protein Ag25784 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction