missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Favezelimab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

£397.00 - £1002.00

Specifications

Antigen Favezelimab
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30282628
GREENER_CHOICE
View Documents
Invitrogen™
MA559635
100 μg
£397.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30284504
View Documents
Invitrogen™
MA559636
1 mg
£1002.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Favezelimab, also known as MK-4280, is a therapeutic monoclonal antibody targeting the lymphocyte-activation gene 3 (LAG-3). This immune checkpoint receptor downregulates T-cell activity, playing a crucial role in immune response regulation. Lymphocyte Activation Gene 3 (LAG3), also known as CD223, is an immune checkpoint receptor encoded by the LAG3 gene located on chromosome 12 (12p13.32). Structurally, LAG3 is closely related to the CD4 molecule, sharing four extracellular immunoglobulin-like domains and a highly conserved cytoplasmic tail, which mediates its inhibitory signaling. Functionally, LAG3 is expressed on activated T cells, natural killer (NK) cells, B cells, and plasmacytoid dendritic cells, playing a crucial role in regulating the immune response. It acts by binding to its primary ligand, MHC class II molecules, and can also interact with other ligands such as fibrinogen-like protein 1 and galectin-3. Through these interactions, LAG3 transmits inhibitory signals to reduce the proliferation and activation of effector T cells, thus maintaining immune homeostasis and preventing autoimmunity. In cancer, LAG3 contributes to the immune evasion of tumors by suppressing anti-tumor T cell activity, making it a significant target for novel cancer immunotherapies.
Specifications

Specifications

Favezelimab
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG4SP
Human
Primary
Protein A
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.