missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FABP1 (L-FABP) Recombinant Protein
Description
This product contains a purified 127 amino acid fragment (aa1-127) of human FABP-L protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.
FABP1 encodes the fatty acid binding protein found in liver. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP1 and FABP6 (the ileal fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism.
Specifications
Specifications
| Accession Number | P07148 |
| Concentration | 0.745 mg/mL |
| For Use With (Application) | Control, Western Blot |
| Formulation | tris HCl with 50% glycerol and no preservative; pH 8 |
| Gene ID (Entrez) | 2168 |
| Molecular Weight (g/mol) | 18.71 kDa |
| Name | Human FABP1 (L-FABP) |
| Quantity | 100 μg |
| Source | E. Coli |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?