missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FATP6/SLC27A6 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-81909CL1
This item is not returnable.
View return policy
Description
FATP6/SLC27A6 Polyclonal antibody specifically detects FATP6/SLC27A6 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| FATP6/SLC27A6 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| ACSVL2VLCSH1, DKFZp779M0564, EC 6.2.1, EC 6.2.1.7, FACVL2hVLCS-H1, FATP1, FATP6, FATP-6, solute carrier family 27 (fatty acid transporter), member 6, Solute carrier family 27 member 6, very long-chain 2, Very long-chain acyl-CoA synthetase homolog 1, VLCS-H1 | |
| SLC27A6 antibody was raised against a 16 amino acid peptide near the amino terminus of human SLC27A6. The immunogen is located within amino acids 180 - 230 of SLC27A6. Amino Acid Squence: KEKLSTSPDEPVPRSH | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 28965 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction