missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FATP5/SLC27A5 Antibody (CL0215) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52880H
This item is not returnable.
View return policy
Description
FATP5/SLC27A5 Monoclonal antibody specifically detects FATP5/SLC27A5 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| FATP5/SLC27A5 | |
| Monoclonal | |
| HRP | |
| ACSBBACS, ACSVL6BAL, BA-CoA ligase, Bile acid-CoA ligase, bile acyl-CoA synthetase, Cholate--CoA ligase, EC 6.2.1.7, FACVL3FATP-5, FATP5Solute carrier family 27 member 5, Fatty acid transport protein 5, Fatty-acid-coenzyme A ligase, very long-chain 3, FLJ22987, solute carrier family 27 (fatty acid transporter), member 5, VLACSRVery long-chain acyl-CoA synthetase homolog 2, VLCSH2Very long-chain acyl-CoA synthetase-related protein, VLCS-H2VLACS-related | |
| This antibody was developed using a recombinant protein derived from Q9Y2P5, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0215 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 10998 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction