missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FATP4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11013-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of a family of fatty acid transport proteins, which are involved in translocation of long-chain fatty acids cross the plasma membrane. This protein is expressed at high levels on the apical side of mature enterocytes in the small intestine, and appears to be the principal fatty acid transporter in enterocytes. Clinical studies suggest this gene as a candidate gene for the insulin resistance syndrome. Mutations in this gene have been associated with ichthyosis prematurity syndrome.Specifications
| FATP4 | |
| Polyclonal | |
| Unconjugated | |
| SLC27A4 | |
| ACSVL4, FATP 4, FATP4, Fatty acid transport protein 4, SLC27A4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10999 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6P1M0 | |
| SLC27A4 | |
| FATP4 Fusion Protein Ag1490 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction