missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FATP3/SLC27A3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99424-100ul
This item is not returnable.
View return policy
Description
FATP3/SLC27A3 Polyclonal antibody specifically detects FATP3/SLC27A3 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| FATP3/SLC27A3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| EC 6.2.1, EC 6.2.1.-, EC 6.2.1.7, FATP-3, FATP3ACSVL3MGC4365, Fatty acid transport protein 3, long-chain fatty acid transport protein 3, solute carrier family 27 (fatty acid transporter), member 3, Solute carrier family 27 member 3, Very long-chain acyl-CoA synthetase homolog 3, VLCS-3 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human FATP3/SLC27A3 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 11000 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction