missing translation for 'onlineSavingsMsg'
Learn More

FATP3/SLC27A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£328.00 - £513.00

Specifications

Antigen FATP3/SLC27A3
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Regulatory Status RUO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18267254
View Documents
Novus Biologicals
NBP2-57049
100 μL
£513.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18613887
View Documents
Novus Biologicals
NBP2-57049-25ul
25 μL
£328.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FATP3/SLC27A3 Polyclonal specifically detects FATP3/SLC27A3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

FATP3/SLC27A3
Unconjugated
RUO
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
11000
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DTWERFVRRFGPLQVLETYGLTEGNVATINYTGQRGAVGRASWLYKHIFPFSLIRYDVTTGEPIRDPQGHCMATSPGEPGLLVAPVSQQSPFLGYAGGPELAQGKLLKDVFR
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Polyclonal
Rabbit
Human
EC 6.2.1, EC 6.2.1.-, EC 6.2.1.7, FATP-3, FATP3ACSVL3MGC4365, Fatty acid transport protein 3, long-chain fatty acid transport protein 3, solute carrier family 27 (fatty acid transporter), member 3, Solute carrier family 27 member 3, Very long-chain acyl-CoA synthetase homolog 3, VLCS-3
SLC27A3
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.