missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FATP2/SLC27A2 Antibody (466106) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4659JF669
This item is not returnable.
View return policy
Description
FATP2/SLC27A2 Monoclonal antibody specifically detects FATP2/SLC27A2 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| FATP2/SLC27A2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| ACSVL1FATP-2, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.3, EC 6.2.1.7, FACVL1very long-chain fatty-acid-coenzyme A ligase 1, FATP2THCA-CoA ligase, Fatty acid transport protein 2, Fatty-acid-coenzyme A ligase, very long-chain 1, hFACVL1, HsT17226, Long-chain-fatty-acid--CoA ligase, solute carrier family 27 (fatty acid transporter), member 2, Solute carrier family 27 member 2, very-long-chain acyl-CoA synthetase, VLACSVery long-chain-fatty-acid-CoA ligase, VLCSvery long-chain acyl-CoA synthetase | |
| HEK293 human embryonic kidney cell line transfected with human FATP2, Met1-Leu620, Accession # O14975 | |
| RUO | |
| 11001 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 466106 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction