missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAT2 Mouse anti-Human, Clone-8C5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54371-100UG
This item is not returnable.
View return policy
Description
FAT2 Monoclonal specifically detects FAT2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| FAT2 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
| protocadherin Fat 2, CDHF8, CDHR9, FAT tumor suppressor homolog 2 (Drosophila), HFAT2, MEGF1 | |
| Mouse | |
| 480 kDa | |
| 100 μg | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| 8C5 | |
| Unconjugated | |
| PBS with No Preservative | |
| FAT2 | |
| Purified recombinant human FAT2 fusion protein | |
| Protein A or G purified | |
| RUO | |
| 2196 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction