missing translation for 'onlineSavingsMsg'
Learn More

FAT1 Mouse anti-Drosophila, Clone: FAT1-3D7/1, Novus Biologicals™

Product Code. 18328141 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328141 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328141 Supplier Novus Biologicals Supplier No. NBP30720620UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FAT1 Monoclonal specifically detects FAT1 in Drosophila samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FAT1
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone FAT1-3D7/1
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila)
Host Species Mouse
Immunogen Cytoplasmic domain of Drosophila FAT1 protein.
Purification Method Protein L purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2195
Target Species Drosophila
Content And Storage Store at 4°C.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.