missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAT1 Antibody (FAT-1 3D7/1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2693
This item is not returnable.
View return policy
Description
FAT1 Monoclonal specifically detects FAT1 in Drosophila samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| FAT1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| FAT1 | |
| Cytoplasmic domain of drosophila Fat | |
| 0.1 mg | |
| Signal Transduction | |
| 2195 | |
| Drosophila | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| FAT-1 3D7/1 | |
| Unconjugated | |
| FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila) | |
| Mouse | |
| Thiophilic Chromatography | |
| RUO | |
| Primary | |
| FAT (Fat-1). Tested only in Drosophila. Expected to cross-react with Rat (85% identity with immunogen) and Chimpanzee (92% identity with immunogen) due to sequence homology. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction