missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAT1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32275DL594
This item is not returnable.
View return policy
Description
FAT1 Polyclonal antibody specifically detects FAT1 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative)
Specifications
| FAT1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation (Negative) | |
| DyLight 594 | |
| FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila) | |
| The immunogen this antibody was made to, maps to a region between residue 4484 to 4534 of human FAT Tumor Suppressor Homolog 1 using the numbering given in entry NP_005236.2 (GeneID 2195). | |
| 0.1 mL | |
| Signal Transduction | |
| 2195 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction