missing translation for 'onlineSavingsMsg'
Learn More

FAT1 Antibody, CoraFluor™ 1, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP2-32275CL1

Product Code. 30141946

  • £821.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

FAT1 Polyclonal antibody specifically detects FAT1 in Human samples. It is validated for Western Blot,Immunoprecipitation (Negative)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FAT1
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Western Blot, Immunoprecipitation (Negative)
CoraFluor 1
FAT tumor suppressor 1, FAT tumor suppressor homolog 1 (Drosophila)
The immunogen this antibody was made to, maps to a region between residue 4484 to 4534 of human FAT Tumor Suppressor Homolog 1 using the numbering given in entry NP_005236.2 (GeneID 2195).
0.1 mL
Signal Transduction
2195
Store at 4°C in the dark. Do not freeze.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.