missing translation for 'onlineSavingsMsg'
Learn More

Fasudil Hydrochloride, MedChemExpress

Product Code. 16430976
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430976 200 mg 200mg
16421556 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16430976 Supplier Medchem Express Supplier No. HY10341

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fasudil (HA-1077; AT877) Hydrochloride is a nonspecific RhoA/ROCK inhibitor and also has inhibitory effect on protein kinases, with an Ki of 0.33 μM for ROCK1, IC50s of 0.158 μM and 4.58 μM, 12.30 μM, 1.650 μM for ROCK2 and PKA, PKC, PKG, respectively. Fasudil Hydrochloride is also a potent Ca2+ channel antagonist and vasodilator.

Fasudil (HA-1077

Specifications

Chemical Name or Material Fasudil Hydrochloride
CAS 105628-07-7
Color White
Purity Grade Notes Research
Molecular Formula C14H18ClN3O2S
Quantity 200 mg
Synonym HA-1077 HydrochlorideAT-877 Hydrochloride
Solubility Information H2O : 55 mg/mL (167.77 mM; Need ultrasonic) ∣DMSO : ≥ 31 mg/mL (94.56 mM)
SMILES O=S(C1=CC=CC2=C1C=CN=C2)(N3CCNCCC3)=O.Cl[H]
Molecular Weight (g/mol) 327.83
Formula Weight 327.83
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.