missing translation for 'onlineSavingsMsg'
Learn More

Fasudil dihydrochloride, 99+%

Product Code. 15413349
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
This item is not returnable. View return policy

Product Code. 15413349

Brand: Thermo Scientific Alfa Aesar J60594.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 203911-27-7 | C14H19Cl2N3O2S | 364.285 g/mol

Fasudil dihydrochloride is used in the inhibition of contractile responses to potassium chloride, phenylephnne (PHE), prostaglandin and hepatic stellate cells spreading. It is also used in the formation of stress fibers and in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fasudil dihydrochloride is used in the inhibition of contractile responses to potassium chloride, phenylephnne (PHE), prostaglandin and hepatic stellate cells spreading. It is also used in the formation of stress fibers and in medicine.

Solubility
Soluble in water and dimethylsulfoxide.

Notes
Incompatible with strong oxidizing agents. Sore in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 203911-27-7
Molecular Formula C14H19Cl2N3O2S
Molecular Weight (g/mol) 364.285
MDL Number MFCD00153805
InChI Key NOXXIYDYFSNHDF-UHFFFAOYSA-N
PubChem CID 16219471
IUPAC Name 5-(1,4-diazepan-1-ylsulfonyl)isoquinoline;dihydrochloride
SMILES C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CN=C3.Cl.Cl

Specifications

Color White
Quantity 250 mg
Merck Index 14,3942
Synonym fasudil dihydrochloride, ha-1077 dihydrochloride, ha 1077, dihydrochloride, 5-1,4-diazepan-1-yl sulfonyl isoquinoline dihydrochloride, 5-isoquinolinesulfonyl homopiperazine, 2hcl, 5-isoquinolinesulfonyl homopiperazine,2hcl, 1-5-isoquinolinylsulfonyl homopiperazine dihydrochloride, 1-5-isoquinoline-sulfonyl-homopiperazine, fasudil and ha 1077, dihydrochloride, ha-1077 dihydrochloride, solid hplc
Solubility Information Soluble in water and dimethylsulfoxide.
Formula Weight 364.29
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Fasudil dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.