missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Fasudil (Dihydrochloride)

Product Code. 17100262
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17100262 100 mg 100mg
17190252 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17100262 Supplier STEMCELL Technologies Supplier No. 73664
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

RHO/ROCK pathway inhibitor; Inhibits ROCK2

  • Fasudil (also known as HA-1077) is a potent inhibitor of Rho-associated coiled-coil containing protein kinase 2 (ROCK2; IC50 = 1.9 μM)
  • Additionally, it inhibits protein kinase C-related kinase 2 (PRK2), mitogen- and stress-activated protein kinase (MSK1), and mitogen-activated protein kinase-activated protein kinase 1b (MAPKAP-K1b) with IC50 values of 4, 5, and 15 μM, respectively (Davies et al.) This product is supplied as the dihydrochloride salt of the molecule

Specifications

CAS 203911-27-7
Synonym HA-1077
Quantity 100 mg
Molecular Formula C14H17N3O2S · 2HCl
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.