Learn More
Thermo Scientific™ FastRuler High Range DNA Ladder, Ready-to-Use, 500-10,000 bp Promotion
Shop All Thermo Scientific ProductsDescription
The FastRuler High Range DNA Ladder is specifically designed for fast sizing and approximate quantification of double-stranded DNA in 48 well (or 96-well) high throughput gels, as well as in conventional agarose gels. The ladder is a mixture of five blunt-ended chromatography-purified individual DNA fragments that are easily resolved in a short separation distance (10 to 20 mm) after an 8 to 14 minutes run on the appropriate agarose gel.
The ladder is especially useful for electrophoresis of PCR products.
Highlights
- Fast separation (8 to 14 min)
- Short separation distance (10 to 20 mm)
- Sharp bands, easy-to-remember fragment sizes and quantities
- Ready-to-use -premixed with loading dye
- Supplied with loading dye solution for sample DNA
Specifications
Specifications
| Concentration | 6X Loading Dye |
| Product Type | DNA Molecular Weight Marker/Ladder |
| Form | Liquid |
| Kit Contents | 6X MassRuler™ DNA Loading Dye, 1mL |
| Quantity | 50 to 333 Applications |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.