Learn More
Thermo Scientific™ FastRuler DNA Ladders, ready-to-use
Product Code. 10669320 Shop All Thermo Scientific Products
Change view
Click to view available options
Size Range:
10 to 200 bp
100 to 5000 bp
50 to 1500 bp
500 to 10000 bp
Unit Size:
1000µL
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Range Unit Size
10669320 500 to 10000 bp Pack of 2
10410280 10 to 200 bp Pack of 2
10528740 50 to 1500 bp Pack of 2
10190740 100 to 5000 bp 1000µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10669320 Supplier Thermo Scientific™ Supplier No. SM1123
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Thermo Scientific FastRuler DNA Ladders are engineered for fast and convenient sizing and approximate quantification of double stranded DNA fragments.
Thermo Scientific FastRuler DNA Ladders is a fast and convenient tool for double-stranded DNA sizing during electrophoresis. It's specifically engineered for rapid sizing and approximate quantification of DNA on both conventional agarose gels and high-throughput E-Gels (in 48- and 96-well formats).
The ladder is a mixture of five blunt-ended chromatography-purified individual DNA fragments. This allows you to achieve precise results faster, saving valuable lab time.
Highlights
  • Variety of available ranges (from ultra-low to high- range)
  • Fast separation (8 to 14 min)
  • Short separation distance (10 to 20 mm)
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Ready-to-use: premixed with 6X MassRuler DNA Loading Dye for direct loading
  • Stable at room temperature for up to 24 months
Sustainability features
Ready-to-use FastRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
TRUSTED_SUSTAINABILITY
Specifications
Gel Type Agarose
Label or Dye Bromophenol Blue
Ready to Load Yes
Size Range 500 to 10000 bp
Gel Compatibility Agarose Gels
Quantity 2 x 500 μL
Product Line FastRuler
Product Type High Range DNA Ladder