missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastRNA™ Win Kit

Product Code. 16552040 Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 Preps
Unit Size:
50 reactions
This item is not returnable. View return policy

Product Code. 16552040

Brand: MP Biomedicals™ 116080050

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For easy and quick isolation of total RNA from a wide variety of plant samples

  • Designed to isolate and purify high quality, total RNA from plant cells and tissues for better RT-PCR results
  • The use of the FastPrep™ instrument combined with unique lysis buffers guarantees an efficient lysis of the starting material and a simultaneous inactivation of endogenous RNases
  • Genomic DNA is separated from the total RNA by binding to specially optimized mineral carrier particles included in the lysis buffer
  • RNA is then mixed with ethanol and transferred to the RNA binding membrane
  • Total RNA then binds, contaminants are washed away, and the pure RNA is eluted
  • No phenol or chloroform is required
  • Includes protocol for simultaneous isolation of proteins
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
For Use With (Application) Northern Blot, RNA Dot Blots, In-Vitro Translation, RT-PCR, ddRT-PCR, cDNA-Libraries, and TaqMan&Trade; Analysis and Array Technologies
No. of Reactions 50
Nucleic Acid Type RNA
Product Type FastRNA™ Win Kit
Quantity 50 Preps
Sample Source Plant Tissues
Sterility Non-sterile

For research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.