missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FASTKD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17464-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that is localized in the mitochondrial inner compartment and that may play a role in mitochondrial apoptosis. Nonsense mutations have been reported to result in cytochrome c oxidase deficiency.Specifications
| FASTKD2 | |
| Polyclonal | |
| Unconjugated | |
| FASTKD2 | |
| FAST kinase domains 2, FASTKD2, KIAA0971 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22868, 301463, 75619 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5M7V7, Q922E6, Q9NYY8 | |
| Fastkd2 | |
| FASTKD2 Fusion Protein Ag11314 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction