missing translation for 'onlineSavingsMsg'
Learn More

FASTKD2 Antibody (1B11), Novus Biologicals™

Product Code. 18361659 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361659 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361659 Supplier Novus Biologicals Supplier No. H00022868M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FASTKD2 Monoclonal antibody specifically detects FASTKD2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FASTKD2
Applications ELISA
Classification Monoclonal
Clone 1B11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055744
Gene Alias FAST kinase domains 2, KIAA0971FAST kinase domain-containing protein 2
Host Species Mouse
Immunogen KIAA0971 (NP_055744, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLTTLKPFGSVSVESKMNNKAGSFFWNLRQFSTLVSTSRTMRLCCLGLCKPKIVHSNWNILNNFHNRMQSTDIIRYLFQDAFIFKSDVGFQTKGISTLTA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 22868
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.