missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FASTK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16434
This item is not returnable.
View return policy
Description
FASTK Polyclonal specifically detects FASTK in Human samples. It is validated for Western Blot.
Specifications
| FASTK | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| EC 2.7.11.8, Fas-activated serine/threonine kinase, FASTFAST kinase | |
| Rabbit | |
| 61 kDa | |
| 0.1 mL | |
| Apoptosis | |
| 10922 | |
| Human | |
| IgG |
| Western Blot | |
| 0.88mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q14296 | |
| FASTK | |
| Recombinant protein encompassing a sequence within the center region of human FAST. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (91%), Dog (96%), Pig (92%), Bovine (90%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction