missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals FastDNA™ SPIN Kit for Soil

Product Code. 17584885 Shop All MP Biomedicals Products
Change view
Click to view available options
No. of Reactions:
100 Preps
50 Preps
Unit Size:
1 set
50 preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
17584885 100 Preps 1 set
11492400 50 Preps 50 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17584885 Supplier MP Biomedicals Supplier No. 116560300
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

FastDNA™ SPIN Kit for Soil is used for rapid isolation of PCR-ready genomic DNA from any environmental sample.

  • The FastDNA™ SPIN Kit for Soil quickly and efficiently isolates bacterial, fungal, plant, and animal genomic DNA from soil and other environmental samples.
  • Soil is processed by a FastPrep™ instrument and Lysing Matrix E tubes designed to efficiently lyse all microorganisms.
  • The released DNA is purified by a silica-based spin filter method and is suitable for PCR analysis and other downstream applications.
  • Complete lysis of all organisms, including historically difficult sources such as eubacterial spores and endospores, gram positive bacteria, yeast, algae, nematodes and fungi is achieved within seconds using a FastPrep instrument and Lysing Matrix E tubes.
  • The FastDNA Spin Kit for Soil delivers the highest DNA yield from any environmental sample including soil, sediments, sludge, compost, manure, rhizosphere or waste water.
  • The FastDNA Spin Kit for Soil contains Lysing Matrix tubes and all the reagents needed to extract DNA in a process that efficiently removes humic acids and other PCR inhibitors.

Specifications

Capacity 2 mL
Concentration Up to 500 mg per prep
Format Kit
Isolation Technology Glass Milk
Sample Type Soil, Feces, Sludge, Wastewater, Environmental, Compost, Manure, Rhizosphere
For Use With (Equipment) FastPrep™ Instrument
For Use With (Application) Isolate bacterial, fungal, plant, and animal genomic DNA from soil and other environmental samples, including sediment, sludge, compost, manure, rhizosphere, waste water and more
No. of Reactions 100 Preps
Product Type Nucleic Acid Isolation, Mini-prep
Species Isolate bacterial, fungal, plant, and animal genomic DNA
Target DNA
Test Time 30 min.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1SET (100PREP) DNA PURIFICATION KIT
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 C
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P404-Store in a closed container.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sodium dodecyl sulphate, guanidine thiocyanate, acetic acid

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.