Learn More
Description
Lambda DNA (dam-), 0.7% agarose, 8 cleavage sites
FastDigest enzymes are an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.
The universal buffer allows rapid single-, double- or multiple DNA digestion within 5-15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. DNA modifying enzymes, such as Klenow Fragment, T4 DNA Ligase, alkaline phosphatases and T4 DNA Polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.
For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.
Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.
Highlights
- 100% activity of all FastDigest enzymes in the universal buffer
- 100% buffer compatibility with downstream applications
- Complete digestion in 5-15 minutes
- Direct loading on gels
- No star activity
- 176 FastDigest enzymes available
Specifications
Specifications
| Components | 10X FastDigest Buffer and 10X FastDigest Green Buffer |
| For Use With (Application) | Fast clone analysis; Fast preparation of DNA for cloning; Digestion of PCR products; Fast RFLP genotyping; Digestion of difficult-to-cleave DNA |
| Quantity | 300 μL |
| Product Type | FastDigest™ BclI |
0
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.