missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fast Skeletal Myosin Heavy Chain 1 Antibody (LM5), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50401AF750
This item is not returnable.
View return policy
Description
Fast Skeletal Myosin Heavy Chain 1 Monoclonal specifically detects Fast Skeletal Myosin Heavy Chain 1 in Chicken samples. It is validated for Western Blot.
Specifications
| Fast Skeletal Myosin Heavy Chain 1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Purified chicken pectorial muscle | |
| 0.1 mL | |
| Chicken | |
| IgG |
| Western Blot | |
| LM5 | |
| Western Blot | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction