missing translation for 'onlineSavingsMsg'
Learn More

Fast Green FCF (Technical Grade), TRC

Product Code. 30396611
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396611 10 g
30399201 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396611 Supplier Toronto Research Chemicals Supplier No. TRCF15000510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2353-45-9
Molecular Formula C37 H34 N2 O10 S3 . 2 Na
Molecular Weight (g/mol) 808.85
Synonym N-Ethyl-N-[4-[[4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl](4-hydroxy-2-sulfophenyl)methylene]-2,5-cyclohexadien-1-ylidene]-3-sulfobenzenemethanaminium inner salt sodium salt (1:2), N-Ethyl-N-[4-[[4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl](4-hydroxy-2-sulfophenyl)methylene]-2,5-cyclohexadien-1-ylidene]-3-sulfobenzenemethanaminium hydroxide inner salt disodium salt, N-Ethyl-N-[4-[[4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl](4-hydroxy-2-sulfophenyl)methylene]-2,5-cyclohexadien-1-ylidene]-3-sulfobenzenemethanaminium inner salt disodium salt, C.I. Food Green 3, C.I. Food Green 3 disodium salt, Fast Green FCF, C.I. 42053, FD&C Green No. 3, FD&C Green No. 3, FD&C Green No. 3, Fast Green, Food Green 3, Food Green No. 3, Green No. 3, Japan Food Green No. 3, Japan Green 3, Japan Green No. 3, NSC 379443
IUPAC Name disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate
SMILES [Na+].[Na+].CCN(Cc1cccc(c1)S(=O)(=O)[O-])c2ccc(cc2)C(=C3C=CC(=[N+](CC)Cc4cccc(c4)S(=O)(=O)[O-])C=C3)c5ccc(O)cc5S(=O)(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/C37H36N2O10S3.2Na/c1-3-38(24-26-7-5-9-33(21-26)50(41,42)43)30-15-11-28(12-16-30)37(35-20-19-32(40)23-36(35)52(47,48)49)29-13-17-31(18-14-29)39(4-2)25-27-8-6-10-34(22-27)51(44,45)46;;/h5-23H,3-4,24-25H2,1-2H3,(H3,41,42,43,44,45,46,47,48,49);;/q;2*+1/p-2
Formula Weight 808.1171
Chemical Name or Material Fast Green FCF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.