missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Green FCF, Electrophoresis Reagent

Product Code. 15447979
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447979 100 g 100g
15437979 25 g 25g
15427979 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15447979 Supplier Thermo Scientific Chemicals Supplier No. J66570.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2353-45-9 | C37H34N2Na2O10S3, C37H34N2Na2O10S3 | 808.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2353-45-9
Molecular Formula C37H34N2Na2O10S3, C37H34N2Na2O10S3
Synonym C.I. 42053

Specifications

Content And Storage Ambient temperatures; Store under Nitrogen
Formula Weight 808.86
Melting Point ≈290°C (dec.)
InChI Key XJBPDZVCYTYRIN-UHFFFAOYSA-L
SMILES [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
IUPAC Name disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium
Molecular Weight (g/mol) 808.84
Quantity 100 g
Sensitivity Hygroscopic
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.