missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Fast Green FCF

Product Code. 16432429
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
16432429 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 16432429

Brand: MedChemExpress HYD0914

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fast Green FCF is a sea green triarylmethane food dye.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Product Type Dye Reagents
Quantity 1 g
Shipping Condition Room temperature
Synonym FD & C Green No. 3 ∣ Food green 3 ∣ C.I. 42053
CAS 2353-45-9
Molecular Weight (g/mol) 808.85 g/mol
Solubility H2O : 10 mg/mL (12.36 mM; Need ultrasonic)
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.