missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Blue BB salt

Product Code. 11382217
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
missing translation for 'unitSize'
1g
25g
5g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 11382217

missing translation for 'mfr': Thermo Scientific Chemicals L09704.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 5486-84-0 | C34H36Cl4N6O6Zn | 831.878 g/mol

Fast Blue BB salt is used as a general substrate for β-glycosidase. Two moles of 6-bromo-2-napthol-β-d-glucopyranoside are cleaved by β-glucosidases into 2 moles of d-glucose and 2 moles of 6-bromo-2-napthol. The 2 moles of 6-bromo-2-napthol formed are coupled with 1 mole of Fast Blue BB salt [4-benzoylamino-2,5-diethoxybenzenediazonium chloride hemi (zinc chloride) salt], resulting in 2 moles of the corresponding dye of 6-bromo-2-naphthol (reddish color). It is a commonly used stain to observe myelin under light microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fast Blue BB salt is used as a general substrate for β-glycosidase. Two moles of 6-bromo-2-napthol-β-d-glucopyranoside are cleaved by β-glucosidases into 2 moles of d-glucose and 2 moles of 6-bromo-2-napthol. The 2 moles of 6-bromo-2-napthol formed are coupled with 1 mole of Fast Blue BB salt [4-benzoylamino-2,5-diethoxybenzenediazonium chloride hemi (zinc chloride) salt], resulting in 2 moles of the corresponding dye of 6-bromo-2-naphthol (reddish color). It is a commonly used stain to observe myelin under light microscopy.

Solubility
Soluble in water.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 5486-84-0
Molecular Formula C34H36Cl4N6O6Zn
Molecular Weight (g/mol) 831.878
MDL Number MFCD00074765
InChI Key CMFRFQODFZBKTI-UHFFFAOYSA-L
Synonym 4-Benzamido-2,5-diethoxybenzene diazonium chloride hemi zinc salt; C.I. 37175
PubChem CID 131675364
IUPAC Name zinc;N-(4-diazonio-2,5-diethoxyphenyl)benzenecarboximidate;dichloride;dihydrochloride
SMILES CCOC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OCC)[N+]#N.CCOC1=CC(=C(C=C1N=C(C2=CC=CC=C2)[O-])OCC)[N+]#N.Cl.Cl.[Cl-].[Cl-].[Zn+2]

Spécification

Melting Point ∼157°C (decomposition)
Beilstein 1845288
Solubility Information Soluble in water.
Formula Weight 831.89
Odor Odorless
Sensitivity Light sensitive
Quantity 25 g
Chemical Name or Material Fast Blue BB salt

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis