missing translation for 'onlineSavingsMsg'
Learn More

Calcium nitride, 99% (metals basis)

Product Code. 11372218
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
This item is not returnable. View return policy

Product Code. 11372218

Brand: Thermo Scientific Alfa Aesar 013124.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12013-82-0 | Ca3N2

Calcium nitride is used in the production of light-emitting diode (LED) phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12013-82-0
Molecular Formula Ca3N2
MDL Number MFCD00015985

Specifications

Melting Point 1195°C
Color Brown
Physical Form Pieces
Quantity 50 g
Assay Percent Range 99% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 148.25
Odor Ammonia-like
Sensitivity Moisture sensitive
Density 2.63 g/cm3
Chemical Name or Material Calcium nitride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.